COM448611769 Combi-Blocks, Inc.

3-(3,5-Dimethyl-4-ethoxyphenyl)aniline

  • MDL number MFCD12913929
  • Cas Number : 1187386-08-8
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1396

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1396
Availability details for
Quantity
  • Availability
  • Add to Cart