COM448611771 Combi-Blocks, Inc.

2-Bromo-5-fluoro-4-thiocyanatoaniline

  • MDL number MFCD11855815
  • Cas Number : 1133115-25-9
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1398
Availability details for
Quantity
  • Availability
  • Add to Cart