COM448611773 Combi-Blocks, Inc.

5-Bromo-2-methyl-4-thiocyanatoaniline

  • MDL number MFCD11855817
  • Cas Number : 1081803-34-0
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1400
Availability details for
Quantity
  • Availability
  • Add to Cart