COM448611783 Combi-Blocks, Inc.

N-BOC-N-isopropyl 4-bromoaniline

  • MDL number MFCD11855823
  • Cas Number : 1133115-32-8
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1412
Availability details for
Quantity
  • Availability
  • Add to Cart