COM448611789 Combi-Blocks, Inc.

4,5-Dichloro-2-methylaniline

  • MDL number MFCD11855825
  • Cas Number : 2387-08-8
  • Molecular Weight : 176.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1418

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1418
Availability details for
Quantity
  • Availability
  • Add to Cart