COM448611790 Combi-Blocks, Inc.

1-Bromo-5-fluoro-4-methyl-2-nitrobenzene

  • MDL number MFCD11042845
  • Cas Number : 1345471-69-3
  • Molecular Weight : 234.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1419

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1419
Availability details for
Quantity
  • Availability
  • Add to Cart