COM448611800 Combi-Blocks, Inc.

4,4'-Diamino-3,3'-difluorobiphenyl

  • MDL number MFCD01658278
  • Cas Number : 448-97-5
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1430
Availability details for
Quantity
  • Availability
  • Add to Cart