COM448611801 Combi-Blocks, Inc.

3-Acetamido-1-bromo-2-nitrobenzene

  • MDL number MFCD11855831
  • Cas Number : 99233-18-8
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1431
Availability details for
Quantity
  • Availability
  • Add to Cart