COM448611802 Combi-Blocks, Inc.

3,5-Dichloro-4-thiocyanatoaniline

  • MDL number MFCD11855832
  • Cas Number : 7494-00-0
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1432
Availability details for
Quantity
  • Availability
  • Add to Cart