COM448611828 Combi-Blocks, Inc.

3-Amino-4-chlorobenzenesulfonic acid

  • MDL number MFCD00035764
  • Cas Number : 98-36-2
  • Molecular Weight : 207.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1474
Availability details for
Quantity
  • Availability
  • Add to Cart