COM448611846 Combi-Blocks, Inc.

1-BOC-4-(4-nitrophenoxy)piperidine

  • MDL number MFCD06200882
  • Cas Number : 138227-62-0
  • Molecular Weight : 322.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1499
Availability details for
Quantity
  • Availability
  • Add to Cart