COM448611858 Combi-Blocks, Inc.

5-Acetamidoisoquinoline

  • MDL number MFCD05981994
  • Cas Number : 27461-33-2
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1515
Availability details for
Quantity
  • Availability
  • Add to Cart