COM448611881 Combi-Blocks, Inc.

1-Benzyloxy-2-fluoro-4-nitrobenzene

  • MDL number MFCD12925758
  • Cas Number : 76243-24-8
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1550

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1550
Availability details for
Quantity
  • Availability
  • Add to Cart