COM448611884 Combi-Blocks, Inc.

Diethyl 2-((2-amino-4-chlorophenylamino)methylene)malonate

  • MDL number MFCD13195638
  • Cas Number : 1199773-10-8
  • Molecular Weight : 312.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1555

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1555
Availability details for
Quantity
  • Availability
  • Add to Cart