COM448611888 Combi-Blocks, Inc.

4-(Benzyloxy)-3-bromo-5-fluoroaniline

  • MDL number MFCD15143448
  • Cas Number : 1215206-01-1
  • Molecular Weight : 296.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1561
Availability details for
Quantity
  • Availability
  • Add to Cart