COM448611896 Combi-Blocks, Inc.

1-Acetamido-4-bromo-2-(trifluoromethoxy)benzene

  • MDL number MFCD00203979
  • Cas Number : 175278-18-9
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1575
Availability details for
Quantity
  • Availability
  • Add to Cart