COM448611899 Combi-Blocks, Inc.

4-Acetamido-2-bromo-5-nitroanisole

  • MDL number MFCD16618927
  • Cas Number : 7357-66-6
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1578
Availability details for
Quantity
  • Availability
  • Add to Cart