COM448611903 Combi-Blocks, Inc.

N-Acetyl 4-bromo-3-chloroaniline

  • MDL number MFCD00040848
  • Cas Number : 22459-81-0
  • Molecular Weight : 248.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1582

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1582
Availability details for
Quantity
  • Availability
  • Add to Cart