COM448611907 Combi-Blocks, Inc.

2-Bromo-1-ethoxy-4-nitrobenzene

  • MDL number MFCD12547883
  • Cas Number : 58244-42-1
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1593
Availability details for
Quantity
  • Availability
  • Add to Cart