COM448611921 Combi-Blocks, Inc.

2-Bromo-4,5-dichloroaniline

  • MDL number MFCD00672938
  • Cas Number : 1940-30-3
  • Molecular Weight : 240.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1608

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1608
Availability details for
Quantity
  • Availability
  • Add to Cart