COM448611944 Combi-Blocks, Inc.

4-Acetamido-3-bromocumene

  • MDL number MFCD00100041
  • Cas Number : 68748-07-2
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2560
Availability details for
Quantity
  • Availability
  • Add to Cart