COM448611955 Combi-Blocks, Inc.

N-Acetyl 2-bromo-4-methoxyaniline

  • MDL number MFCD11110217
  • Cas Number : 79069-37-7
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2576

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2576
Availability details for
Quantity
  • Availability
  • Add to Cart