COM448611958 Combi-Blocks, Inc.

tert-butyl N-(4-bromo-2-methoxyphenyl)carbamate

  • MDL number MFCD03426369
  • Cas Number : 262433-01-2
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2586

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2586
Availability details for
Quantity
  • Availability
  • Add to Cart