COM448611961 Combi-Blocks, Inc.

N-Boc 2-Iodo-4-fluoroaniline

  • MDL number MFCD13189513
  • Cas Number : 1060813-09-3
  • Molecular Weight : 337.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2589
Availability details for
Quantity
  • Availability
  • Add to Cart