COM448611968 Combi-Blocks, Inc.

5-Benzyloxy-4-bromo-2-nitroaniline

  • MDL number MFCD17214241
  • Cas Number : 1255574-69-6
  • Molecular Weight : 323.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2598
Availability details for
Quantity
  • Availability
  • Add to Cart