COM448611991 Combi-Blocks, Inc.

4-Methoxy-3-nitrobenzonitrile

  • MDL number MFCD00221439
  • Cas Number : 33224-23-6
  • Molecular Weight : 178.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2640
Availability details for
Quantity
  • Availability
  • Add to Cart