COM448612000 Combi-Blocks, Inc.

2-(Benzyloxy)-4-bromo-1-nitrobenzene

  • MDL number MFCD28015519
  • Cas Number : 849210-10-2
  • Molecular Weight : 308.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2662
Availability details for
Quantity
  • Availability
  • Add to Cart