COM448612005 Combi-Blocks, Inc.

2-Bromo-4-chloro-6-methoxyaniline

  • MDL number MFCD18311825
  • Cas Number : 1261895-84-4
  • Molecular Weight : 236.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2668

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2668
Availability details for
Quantity
  • Availability
  • Add to Cart