COM448612009 Combi-Blocks, Inc.

4-Bromo-5-ethoxy-2-nitro-N-propylaniline

  • MDL number MFCD18783107
  • Cas Number : 1280786-62-0
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2672
Availability details for
Quantity
  • Availability
  • Add to Cart