COM448612013 Combi-Blocks, Inc.

4-Bromo-N-butyl-5-methoxy-2-nitroaniline

  • MDL number MFCD18783110
  • Cas Number : 1280786-70-0
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2676

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2676
Availability details for
Quantity
  • Availability
  • Add to Cart