COM448612014 Combi-Blocks, Inc.

4-Bromo-N-butyl-5-ethoxy-2-nitroaniline

  • MDL number MFCD18783111
  • Cas Number : 1280786-89-1
  • Molecular Weight : 317.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2677
Availability details for
Quantity
  • Availability
  • Add to Cart