COM448612027 Combi-Blocks, Inc.

1,5-Difluoro-2-methyl-4-nitrobenzene

  • MDL number MFCD16620288
  • Cas Number : 179011-38-2
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2694
Availability details for
Quantity
  • Availability
  • Add to Cart