COM448612033 Combi-Blocks, Inc.

4-Bromo-2-nitro-5-propoxy-N-propylaniline

  • MDL number MFCD19237157
  • Cas Number : 1311197-83-7
  • Molecular Weight : 317.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2705

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2705
Availability details for
Quantity
  • Availability
  • Add to Cart