COM448612034 Combi-Blocks, Inc.

tert-Butyl N-(4-bromo-2,6-dimethylphenyl)carbamate

  • MDL number MFCD08703147
  • Cas Number : 1365988-24-4
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2707
Availability details for
Quantity
  • Availability
  • Add to Cart