COM448612035 Combi-Blocks, Inc.

3-Bromo-6-nitrodurene

  • MDL number MFCD01463901
  • Cas Number : 52415-10-8
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2708

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2708
Availability details for
Quantity
  • Availability
  • Add to Cart