COM448612037 Combi-Blocks, Inc.

4-Bromo-2-chloro-5-nitroaniline

  • MDL number MFCD18783113
  • Cas Number : 872820-00-3
  • Molecular Weight : 251.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2710

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2710
Availability details for
Quantity
  • Availability
  • Add to Cart