COM448612060 Combi-Blocks, Inc.

3,5-Dimethyl-4-(2-nitrophenyl)-1,2-oxazole

  • MDL number MFCD22205726
  • Cas Number : 136295-82-4
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2747
Availability details for
Quantity
  • Availability
  • Add to Cart