COM448612061 Combi-Blocks, Inc.

4-(4-BOC-piperazino)-3-bromoaniline

  • MDL number MFCD19684092
  • Cas Number : 1314985-71-1
  • Molecular Weight : 356.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2748

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2748
Availability details for
Quantity
  • Availability
  • Add to Cart