COM448612068 Combi-Blocks, Inc.

4-Bromo-6-fluorobenzene-1,3-diamine

  • MDL number MFCD19684093
  • Cas Number : 166818-71-9
  • Molecular Weight : 205.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2760
Availability details for
Quantity
  • Availability
  • Add to Cart