COM448612071 Combi-Blocks, Inc.

2-Bromo-4-(4-methoxyphenyl)methyl-6-nitroaniline

  • MDL number MFCD19684094
  • Cas Number : 1314987-29-5
  • Molecular Weight : 337.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2764
Availability details for
Quantity
  • Availability
  • Add to Cart