COM448612075 Combi-Blocks, Inc.

3-Amino-4-(trifluoromethoxy)benzonitrile

  • MDL number MFCD09745166
  • Cas Number : 1220630-86-3
  • Molecular Weight : 202.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2768

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2768
Availability details for
Quantity
  • Availability
  • Add to Cart