COM448612084 Combi-Blocks, Inc.

3,4-Difluoro-2-hydroxyaniline, HCl

  • MDL number MFCD19981504
  • Cas Number : 1330750-27-0
  • Molecular Weight : 181.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2782

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2782
Availability details for
Quantity
  • Availability
  • Add to Cart