COM448612085 Combi-Blocks, Inc.

3-Nitro-4-(trifluoromethoxy)benzamide

  • MDL number MFCD13185754
  • Cas Number : 1214323-50-8
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2783

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2783
Availability details for
Quantity
  • Availability
  • Add to Cart