COM448612094 Combi-Blocks, Inc.

4-Acetamido-3-hydroxybenzoic acid

  • MDL number MFCD00869774
  • Cas Number : 10098-40-5
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2797
Availability details for
Quantity
  • Availability
  • Add to Cart