COM448612100 Combi-Blocks, Inc.

N-(2,2,2-Trifluoroethyl) 4-nitrobenzamide

  • MDL number MFCD07850267
  • Cas Number : 365425-94-1
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2809
Availability details for
Quantity
  • Availability
  • Add to Cart