COM448612114 Combi-Blocks, Inc.

Dimethyl 2-(3-fluoro-2-nitrophenyl)malonate

  • MDL number MFCD20441782
  • Cas Number : 872141-24-7
  • Molecular Weight : 271.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2838
Availability details for
Quantity
  • Availability
  • Add to Cart