COM448612119 Combi-Blocks, Inc.

5-Bromo-2-fluoro-3-nitrotoluene

  • MDL number MFCD22205727
  • Cas Number : 1375068-74-8
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2843
Availability details for
Quantity
  • Availability
  • Add to Cart