COM448612129 Combi-Blocks, Inc.

N-Cyclohexyl-5-fluoro-2-nitroaniline

  • MDL number MFCD17336278
  • Cas Number : 1270967-29-7
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2863
Availability details for
Quantity
  • Availability
  • Add to Cart