COM448612158 Combi-Blocks, Inc.

Benzyl N-(3-bromo-2-fluorophenyl)carbamate

  • MDL number MFCD20529437
  • Cas Number : 903556-49-0
  • Molecular Weight : 324.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2899
Availability details for
Quantity
  • Availability
  • Add to Cart