COM448612163 Combi-Blocks, Inc.

4-Bromo-N-cyclohexyl-2,3-difluoro-6-nitroaniline

  • MDL number MFCD21609498
  • Cas Number : 1365272-99-6
  • Molecular Weight : 335.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2910
Availability details for
Quantity
  • Availability
  • Add to Cart